4'-(Methylthio)acetophenone ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L18074.14 L18074.06
L18074.14EA 184 GBP
L18074.14 L18074.06
4'-(Methylthio)acetophenone ≥99%
4'-(Methylthio)acetophenone

Purity: 99% CAS No.: 1778-09-2 Molecular Formula: C9H10OS Formula Weight: 166.24 Melting Point: 80-82° Boiling Point: 135°/1mm MDL No.: MFCD00039835

Formula: C₉H₁₀OS
Boiling Pt: 135 °C (1 mmHg)
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00039835
CAS Number: 1778-09-2
EINECS: 217-213-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR