(3,3,3-Trifluoropropyl)dimethoxymethylsilane ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane, 1,1,1-Trifluoro-3-[dimethoxy(methyl)silyl]propane

L17292.06 L17292.14
L17292.06EA 41.7 GBP
L17292.06 L17292.14
(3,3,3-Trifluoropropyl)dimethoxymethylsilane ≥97%
(3,3,3-Trifluoropropyl)dimethoxymethylsilane

Purity: 97% CAS No.: 358-67-8 Molecular Formula: C6H13F3O2Si Formula Weight: 202.25 Flash Point: 58°(136°F) Refractive Index: 1.3576 Boiling Point: 85°/150mm Beilstein Reference: 1753237 UN No.: 1993 MDL No.: MFCD00053175

Formula: C₆H₁₃F₃O₂Si
Boiling Pt: 85 °C (150 mmHg)
Density: 1.100
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053175
CAS Number: 358-67-8
EINECS: 206-619-4
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR