2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine Mass Spec Std

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2,4,6-Tris(perfluoropropyl)-1,3,5-triazine

L16883.06 L16883.03
L16883.06EA 121 GBP
L16883.06 L16883.03
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine Mass Spec Std
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine

CAS No.: 915-76-4 Molecular Formula: C12F21N3 Formula Weight: 585.12 Refractive Index: 1.3110 Boiling Point: 165° Beilstein Reference: 381010 MDL No.: MFCD00042438

Formula: C₁₂F₂₁N₃
MW: 585.12 g/mol
Boiling Pt: 165 °C
Density: 1.716
Storage Temperature: Ambient
MDL Number: MFCD00042438
CAS Number: 915-76-4
EINECS: 213-023-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR