Amberlite® IR-120 (Na) ion exchange resin

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L15199.22 L15199.36 L15199.0E
L15199.22EA 20 GBP
L15199.22 L15199.36 L15199.0E
Amberlite® IR-120 (Na) ion exchange resin
Amberlite® IR-120 (Na)

CAS No.: 78922-04-0 MDL No.: MFCD00132707

Storage Temperature: Ambient
MDL Number: MFCD00132707
CAS Number: 9002-23-7
Merck Index: 14,382

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR