(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (R)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole

L14582.AC L14582.AA L14582.XF
L14582.ACEA 200 GBP
L14582.AC L14582.AA L14582.XF
(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
(R)-(+)-2-Methyl-CBS-oxazaborolidine

CAS No.: 112022-83-0 Molecular Formula: C18H20BNO Formula Weight: 277.17 Flash Point: 4°(39°F) UN No.: 1993 MDL No.: MFCD00078440

Formula: C₁₈H₂₀BNO
Density: 0.925
Flash Pt: 4 °C (39 °F)
Storage Temperature: Ambient
MDL Number: MFCD00078440
CAS Number: 112022-83-0
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR