Tetrapropylammonium hydroxide 1 M in aqueous solution

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: IPC-TPA-OH

L14328.14 L14328.22 L14328.36
L14328.14EA 24.7 GBP
L14328.14 L14328.22 L14328.36
Tetrapropylammonium hydroxide 1 M in aqueous solution
Tetrapropylammonium hydroxide

CAS No.: 4499-86-9 Molecular Formula: (CH3CH2CH2)4NOH Formula Weight: 203.37 Beilstein Reference: 4207983 UN No.: 3267 MDL No.: MFCD00009360

Formula: C₁₂H₂₉NO
Density: 1.00
Storage Temperature: Ambient
MDL Number: MFCD00009360
CAS Number: 4499-86-9
EINECS: 224-800-6
UN: 3267
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR