(S)-(+)-2-Butanol ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (S)-(+)-sec-Butyl alcohol, (S)-Butan-2-ol, (S)-(+)-sec-Butanol, (S)-(+)-Ethyl methyl carbinol, (S)-(+)-2-Hydroxy butane, (S)-(+)-Methyl ethyl carbinol

L14164.03 L14164.06
L14164.03EA 50 GBP
L14164.03 L14164.06
(S)-(+)-2-Butanol ≥98%
(S)-(+)-2-Butanol

Purity: 98+% CAS No.: 4221-99-2 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 26°(79°F) Refractive Index: 1.3980 Odour: Alcohol-like Boiling Point: 99-100° Beilstein Reference: 1718763 Merck Reference: 14,1541 Optical Rotation: +13.5° (c=10 in methanol) UN No.: 1120 MDL No.: MFCD00064281

Formula: C₄H₁₀O
MW: 74.12 g/mol
Boiling Pt: 99…100 °C
Melting Pt: –114 °C
Density: 0.806
Flash Pt: 26 °C (79 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064281
CAS Number: 4221-99-2
EINECS: 224-168-1
UN: 1120
ADR: 3 III
Merck Index: 13,01540

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR