Dowex® 50WX2 (H) 100-200 mesh

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L13944.18 L13944.30 L13944.0B
L13944.18EA 44.1 GBP
L13944.18 L13944.30 L13944.0B
Dowex® 50WX2 (H) 100-200 mesh
Dowex® 50WX2 (H)

CAS No.: 12612-37-2 MDL No.: MFCD00132722

Storage Temperature: Ambient
MDL Number: MFCD00132722
CAS Number: 12612-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR