Hydrogen peroxide 27% (w/w) in aqueous solution stabilised

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

L13235.AE L13235.AP
L13235.AEEA 20.1 GBP
L13235.AE L13235.AP
Hydrogen peroxide 27% (w/w) in aqueous solution stabilised
Hydrogen peroxide

CAS No.: 7722-84-1 Molecular Formula: H2O2 Formula Weight: 34.02 Refractive Index: 1.3350 Melting Point: -26° Boiling Point: 107° Merck Reference: 14,4798 UN No.: 2014 MDL No.: MFCD00011333

Formula: H₂O₂
Boiling Pt: 107 °C
Melting Pt: –26 °C
Density: 1.100
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II
Merck Index: 14,4798

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR