Methyl ethyl ketone ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

L13185.AP L13185.AU L13185.0F
L13185.APEA 19.9 GBP
L13185.AP L13185.AU L13185.0F
Methyl ethyl ketone ≥99%
Methyl ethyl ketone

Purity: 99% CAS No.: 78-93-3 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -3°(26°F) Refractive Index: 1.3785 Odour: Characteristic Melting Point: -87° Boiling Point: 80° Beilstein Reference: 741880 Merck Reference: 14,6072 UN No.: 1193 MDL No.: MFCD00011648

Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 80 °C
Melting Pt: –87 °C
Density: 0.805
Flash Pt: –3 °C (26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR