Dinonyl sulphide ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L10638.06
L10638.06EA 25.5 GBP
L10638.06
Dinonyl sulphide ≥98%
Dinonyl sulphide

Purity: 98% CAS No.: 929-98-6 Molecular Formula: C18H38S Formula Weight: 286.56 Flash Point: 110°(230°F) Refractive Index: 1.4640 Odour: Unpleasant Melting Point: 15° Boiling Point: 165-167°/3mm Beilstein Reference: 1755874 MDL No.: MFCD00027342

Formula: C₁₈H₃₈S
Boiling Pt: 165…167 °C (3 mmHg)
Melting Pt: 15 °C
Density: 0.855
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00027342
CAS Number: 929-98-6
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR