Hydriodic acid 57% w/w aqueous solution Stabilised

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Hydroiodic acid, Hydrogen iodide, Hydrogen iodide solution

Stabilisation: 1.5% hypophosphorous acid

L10410.22 L10410.36
L10410.22EA 39.5 GBP
L10410.22 L10410.36
Hydriodic acid 57% w/w aqueous solution Stabilised
Hydriodic acid

CAS No.: 10034-85-2 Molecular Formula: HI Formula Weight: 127.91 Physical Form: Liquid Density: 1.701 Sensitivity: Light Sensitive Appearance: Colourless to yellow Odour: Pungent Boiling Point: 126-127° Merck Reference: 14,4776 UN No.: 1787 MDL No.: MFCD00011347

Formula: HI
MW: 127.91 g/mol
Boiling Pt: 126…127 °C
Density: 1.701
Storage Temperature: Ambient
MDL Number: MFCD00011347
CAS Number: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8 II
Merck Index: 14,4776

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR