1-Bromo-4,4,4-trifluorobutane ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

L10237.03 L10237.06
L10237.03EA 27.6 GBP
L10237.03 L10237.06
1-Bromo-4,4,4-trifluorobutane ≥98%
1-Bromo-4,4,4-trifluorobutane

Purity: 98% CAS No.: 406-81-5 Molecular Formula: CF3CH2CH2CH2Br Formula Weight: 191.00 Refractive Index: 1.3820 Boiling Point: 103-105° MDL No.: MFCD00066335

Formula: C₄H₆BrF₃
Boiling Pt: 103…105 °C
Density: 1.555
Storage Temperature: Ambient
MDL Number: MFCD00066335
CAS Number: 406-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR