Isobornyl acrylate ≥85% stabilised, tech.

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (1R,2R,4R)-rel-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl acrylate, Acrylic acid Isobornyl Ester

Stabilisation: Stabilised with 4-methoxyphenol (MEHQ) 100 ppm

L09977.30
L09977.30EA 51.3 GBP
L09977.30
Isobornyl acrylate ≥85% stabilised, tech.
Isobornyl acrylate

Stab. with 100ppm 4-methoxyphenol


Purity: 85% CAS No.: 5888-33-5 Molecular Formula: C13H20O2 Formula Weight: 208.30 Flash Point: 108°(226°F) Refractive Index: 1.4795 Odour: Characteristic Melting Point: -35° Boiling Point: 104-106°/4.5mm Beilstein Reference: 2254825 UN No.: UN3082 MDL No.: MFCD00080424

Formula: C₁₃H₂₀O₂
MW: 208.30 g/mol
Boiling Pt: 104…106 °C (4.5 mmHg)
Melting Pt: < –35 °C
Density: 0.970
Flash Pt: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00080424
CAS Number: 5888-33-5
EINECS: 227-561-6
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR