trans-2-Butenyl acetate 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

L09734.06 L09734.14
L09734.06EA 52.9 GBP
L09734.06 L09734.14
trans-2-Butenyl acetate 95%
trans-2-Butenyl acetate

Purity: 95% CAS No.: 628-08-0 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 31°(87°F) Refractive Index: 1.4200 Boiling Point: 128-130° Beilstein Reference: 1720994 UN No.: 3272 MDL No.: MFCD00039912

Formula: C₆H₁₀O₂
Boiling Pt: 128…130 °C
Density: 0.919
Flash Pt: 31 °C (87 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039912
CAS Number: 628-08-0
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR