Isopropyl acetate 99

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Acetic acid isopropyl ester

L09337.AK L09337.AU L09337.0F
L09337.AKEA 21.7 GBP
L09337.AK L09337.AU L09337.0F
Isopropyl acetate 99
Isopropyl acetate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 87…89 °C
Melting Pt: –73 °C
Density: 0.870
Flash Pt: 4 °C (39 °F)
MDL Number: MFCD00008877
CAS Number: 108-21-4
UN: 1220
ADR: 3 II
Merck Index: 14,5205

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR