1,2-Diphenylcyclopropane cis- and trans-mixture 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L08833.09 L08833.04
L08833.09EA 276 GBP
L08833.09 L08833.04
1,2-Diphenylcyclopropane cis- and trans-mixture 97%
1,2-Diphenylcyclopropane

Purity: 97% CAS No.: 29881-14-9 Molecular Formula: C15H14 Formula Weight: 194.28 Refractive Index: 1.5950 Boiling Point: 175-177°/14mm Beilstein Reference: 2043299 MDL No.: MFCD00040875

Formula: C₁₅H₁₄
Boiling Pt: 175…177 °C (14 mmHg)
Density: 1.020
Storage Temperature: Ambient
MDL Number: MFCD00040875
CAS Number: 29881-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR