Sodium bis(trimethylsilyl)amide 1 M in THF

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: HMDS sodium salt, N-Sodiohexamethyldisilazane, Sodium bis(trimethylsilyl)amide, HMDS-Na

L08779.AE L08779.AP
L08779.AEEA 52 GBP
L08779.AE L08779.AP
Sodium bis(trimethylsilyl)amide 1 M in THF
Sodium bis(trimethylsilyl)amide

CAS No.: 1070-89-9 Molecular Formula: C6H18NNaSi2 Formula Weight: 183.38 Flash Point: -17°(1°F) Beilstein Reference: 3629917 UN No.: 2924 MDL No.: MFCD00009835

Formula: C₆H₁₈NNaSi₂
MW: 183.38 g/mol
Density: 0.904
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009835
CAS Number: 1070-89-9
EINECS: 213-983-8
UN: 2924
ADR: 3 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR