Octyltrimethylammonium bromide 97

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Trimethyl-1-octylammonium bromide

L08610.06 L08610.14 L08610.22
L08610.06EA 25.4 GBP
L08610.06 L08610.14 L08610.22
Octyltrimethylammonium bromide 97
Octyltrimethylammonium bromide

Purity: 97% CAS No.: 2083-68-3 Molecular Formula: CH3(CH2)7N(CH3)3Br Formula Weight: 252.24 Melting Point: 220-222° Beilstein Reference: 3628448

Formula: C₁₁H₂₆BrN
MW: 252.24 g/mol
Melting Pt: 220…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00051873
CAS Number: 2083-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR