4-Nitro-3-(trifluoromethyl)phenol ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

L08228.03 L08228.06
L08228.03EA 20 GBP
L08228.03 L08228.06
4-Nitro-3-(trifluoromethyl)phenol ≥97%
4-Nitro-3-(trifluoromethyl)phenol

Purity: 97% CAS No.: 88-30-2 Molecular Formula: C7H4F3NO3 Formula Weight: 207.11 Melting Point: 74-77° Boiling Point: 135-138°/0.01mm Merck Reference: 14,9253 UN No.: 2811 MDL No.: MFCD00014714

Formula: C₇H₄F₃NO₃
Boiling Pt: 135…138 °C (0.01 mmHg)
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00014714
CAS Number: 88-30-2
EINECS: 201-818-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,9253

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR