7-Azaindole ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L07983.03 L07983.06 L07983.14
L07983.03EA 21.8 GBP
L07983.03 L07983.06 L07983.14
7-Azaindole ≥98%
7-Azaindole

Purity: 98% CAS No.: 271-63-6 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 103-107° Boiling Point: 270° MDL No.: MFCD00005606

Formula: C₇H₆N₂
Boiling Pt: 270 °C
Melting Pt: 103…107 °C
Density: 1.301 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005606
CAS Number: 271-63-6
EINECS: 205-981-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR