3,4-Dimethyl-o-phenylenediamine 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Amino-3,4-dimethylphenylamine, 3,4-Dimethyl-1,2-phenylenediamine, 3,4-Dimethylbenzene-1,2-diamine, 1,2-Diamino-3,4-dimethylbenzene

L07455.03 L07455.06
L07455.03EA 37 GBP
L07455.03 L07455.06
3,4-Dimethyl-o-phenylenediamine 98%
3,4-Dimethyl-o-phenylenediamine

Purity: 98% CAS No.: 41927-01-9 Molecular Formula: C8H12N2 Formula Weight: 136.20 Melting Point: 86-90° Beilstein Reference: 2716459 UN No.: 2811 MDL No.: MFCD00051709

Formula: C₈H₁₂N₂
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00051709
CAS Number: 41927-01-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR