(Chloromethyl)cyclopropane ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Cyclopropyl methyl chloride

L06986.06 L06986.14
L06986.06EA 40.7 GBP
L06986.06 L06986.14
(Chloromethyl)cyclopropane ≥97%
(Chloromethyl)cyclopropane

Purity: 97% CAS No.: 5911-08-0 Molecular Formula: C4H7Cl Formula Weight: 90.55 Storage Temperature: 2°C to 8°C Flash Point: -1°(30°F) Refractive Index: 1.4360 Odour: Recognizable Melting Point: -91° Boiling Point: 87-89° Beilstein Reference: 635697 UN No.: 1993 MDL No.: MFCD00001307

Formula: C₄H₇Cl
MW: 90.55 g/mol
Boiling Pt: 87…89 °C
Melting Pt: –91 °C
Density: 0.980
Flash Pt: –1 °C (30 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001307
CAS Number: 5911-08-0
EINECS: 227-632-1
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR