6-Nitro-o-toluic acid 98

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Methyl-6-nitrobenzoic acid

L06563.06 L06563.14
L06563.06EA 45.3 GBP
L06563.06 L06563.14
6-Nitro-o-toluic acid 98
6-Nitro-o-toluic acid

Purity: 98% CAS No.: 13506-76-8 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 153-156° Beilstein Reference: 2050804

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 353
Melting Pt: 153…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007267
CAS Number: 13506-76-8
EINECS: 236-833-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR