Ethylbenzene 99

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Phenylethane

L05908.AE L05908.AP L05908.0F
L05908.AEEA 22.6 GBP
L05908.AE L05908.AP L05908.0F
Ethylbenzene 99
Ethylbenzene

Purity: 99% CAS No.: 100-41-4 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 15°(59°F) Refractive Index: 1.4950 Odour: Petrol-like Melting Point: -95° Boiling Point: 135-136° Beilstein Reference: 1901871 Merck Reference: 14,3765 UN No.: 1175 MDL No.: MFCD00011647

Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 135…136 °C
Melting Pt: –95 °C
Density: 0.867
Flash Pt: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS Number: 100-41-4
EINECS: 202-849-4
UN: 1175
ADR: 3 II
Merck Index: 13,03801
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67d12bb66494ff09657a029fa1e68b09af34358&vid=3e5678dd6a0342c5ec35b93e48587e8125977cf7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR