2-(4-Fluorophenyl)ethanol ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

L05618.06 L05618.14
L05618.06EA 36.8 GBP
L05618.06 L05618.14
2-(4-Fluorophenyl)ethanol ≥97%
2-(4-Fluorophenyl)ethanol

Purity: 97% CAS No.: 7589-27-7 Molecular Formula: C8H9FO Formula Weight: 140.16 Flash Point: 104°(219°F) Refractive Index: 1.5065 Boiling Point: 118°/20mm Beilstein Reference: 2354310 MDL No.: MFCD00002898

Formula: C₈H₉FO
Boiling Pt: 118 °C (20 mmHg)
Density: 1.121
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002898
CAS Number: 7589-27-7
EINECS: 231-499-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR