L-Adrenaline ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

L04911.06 L04911.14
L04911.06EA 66.8 GBP
L04911.06 L04911.14
L-Adrenaline ≥98%
L-Adrenaline

Purity: 98+% CAS No.: 51-43-4 Molecular Formula: C9H13NO3 Formula Weight: 183.20 Storage Temperature: 2°C to 8°C Melting Point: 216-219° Beilstein Reference: 2368277 Optical Rotation: -52° (c=1 in 0.6N HCl) UN No.: UN2811 MDL No.: MFCD00002204

Formula: C₉H₁₃NO₃
Melting Pt: 216…219 °C
MDL Number: MFCD00002204
CAS Number: 51-43-4
EINECS: 200-098-7
ADR: 6.1,II
Merck Index: 14,3619

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR