Isobutyric acid ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Methylpropionic acid, 2-Methylpropanoic acid

L04038.AE L04038.AP L04038.0F
L04038.AEEA 19.6 GBP
L04038.AE L04038.AP L04038.0F
Isobutyric acid ≥99%
Isobutyric acid

Purity: 99% CAS No.: 79-31-2 Molecular Formula: (CH3)2CHCO2H Formula Weight: 88.11 Flash Point: 59°(138°F) Refractive Index: 1.3928 Odour: Pungent Melting Point: -47° Boiling Point: 153-154° Beilstein Reference: 635770 Merck Reference: 14,5155 UN No.: 2529 MDL No.: MFCD00002658

Formula: C₄H₈O₂
Boiling Pt: 153…154 °C
Melting Pt: –47 °C
Density: 0.948
Flash Pt: 59 °C (138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002658
CAS Number: 79-31-2
EINECS: 201-195-7
UN: 2529
ADR: 3,III
Merck Index: 14,5155

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR