3-(Trifluoromethyl)crotonic acid 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

L03816.06 L03816.14
L03816.06EA 69.8 GBP
L03816.06 L03816.14
3-(Trifluoromethyl)crotonic acid 96%
3-(Trifluoromethyl)crotonic acid

Purity: 96% CAS No.: 69056-67-3 Molecular Formula: C5H5F3O2 Formula Weight: 154.09 Flash Point: 79°(174°F) Refractive Index: 1.3850 Melting Point: 28-30° Boiling Point: 158-163° Beilstein Reference: 1769204 UN No.: UN3261 MDL No.: MFCD00014374

Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 158…163 °C
Melting Pt: 28…30 °C
Density: 1.318
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014374
CAS Number: 69056-67-3
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR