Estradiol, (max. 3% H₂O) 99% (dry weight)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

L03801.03 L03801.06 L03801.14
L03801.03EA 29.9 GBP
L03801.03 L03801.06 L03801.14
Estradiol, (max. 3% H₂O) 99% (dry weight)
Estradiol

Purity: 99% CAS No.: 50-28-2 Molecular Formula: C18H24O2 Formula Weight: 272.39 Melting Point: 175-178° Beilstein Reference: 1914275 Merck Reference: 14,3703 Optical Rotation: +79° (c=1 in dioxane) MDL No.: MFCD00003693

Formula: C₁₈H₂₄O₂
Melting Pt: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00003693
CAS Number: 50-28-2
EINECS: 200-023-8
Merck Index: 14,3703

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR