1-(4-Methoxyphenyl)piperazine dihydrochloride ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

L02007.06
L02007.06EA 0 GBP
L02007.06
1-(4-Methoxyphenyl)piperazine dihydrochloride ≥97%
1-(4-Methoxyphenyl)piperazine dihydrochloride

Purity: 97% CAS No.: 38869-47-5 Molecular Formula: C11H16N2O.2HCl Formula Weight: 265.18 Melting Point: ca 240° dec. Beilstein Reference: 4275784

Formula: C₁₁H₁₈Cl₂N₂O
Melting Pt: ∼ 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012767
CAS Number: 38869-47-5
EINECS: 254-166-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR