5,6-Dihydrouracil 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L01918.03 L01918.06
L01918.03EA 32.4 GBP
L01918.03 L01918.06
5,6-Dihydrouracil 97%
5,6-Dihydrouracil

Purity: 97% CAS No.: 504-07-4 Molecular Formula: C4H6N2O2 Formula Weight: 114.10 Melting Point: 278-280° Beilstein Reference: 112496 MDL No.: MFCD00006029

Formula: C₄H₆N₂O₂
Melting Pt: 278…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00006029
CAS Number: 504-07-4
EINECS: 207-982-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR