2,4-Dichlorophenylboronic acid ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Dihydroxy(2,4-dichlorophenyl)borane, 2,4-Dichlorobenzeneboronic acid, (2,4-Dichlorophenyl)metaboric acid

L01563.03 L01563.06
L01563.03EA 19.2 GBP
L01563.03 L01563.06
2,4-Dichlorophenylboronic acid ≥98%
2,4-Dichlorophenylboronic acid

Purity: 98+% CAS No.: 68716-47-2 Molecular Formula: C6H5BCl2O2 Formula Weight: 190.82 Melting Point: 242-245° Beilstein Reference: 4983878

Formula: C₆H₅BCl₂O₂
Melting Pt: 242…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00013930
CAS Number: 68716-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR