m-Toluic hydrazide ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: m-Toluic acid hydrazide, 3-Methylbenzoic hydrazide

L01043.06 L01043.14
L01043.06EA 24.8 GBP
L01043.06 L01043.14
m-Toluic hydrazide ≥97%
m-Toluic hydrazide

Purity: 97% CAS No.: 13050-47-0 Molecular Formula: C8H10N2O Formula Weight: 150.18 Melting Point: 94-98° Beilstein Reference: 2045125

Formula: C₈H₁₀N₂O
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00014760
CAS Number: 13050-47-0
EINECS: 235-926-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR