Ethyl-O-toluate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Ethyl-2-methylbenzoate, 2-Toluic acid ethyl ester, o-Toluic acid ethyl ester, Ethyl 2-methylbenzoate, 2-Methylbenzoic acid ethyl ester

L00598.14 L00598.22
L00598.14EA 29.1 GBP
L00598.14 L00598.22
Ethyl-O-toluate ≥98%
Ethyl o-toluate

Purity: 98+% CAS No.: 87-24-1 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 91°(195°F) Refractive Index: 1.5070 Boiling Point: 227° Beilstein Reference: 2042517 MDL No.: MFCD00009112

Formula: C₁₀H₁₂O₂
Boiling Pt: 227 °C
Melting Pt: –10 °C
Density: 1.033
Flash Pt: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009112
CAS Number: 87-24-1
EINECS: 201-734-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR