ABTS® Peroxidase Substrate Kit, 1-Component Microwell
Supplier: SERACARE LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5120-0043
KPLI5120-0043EA 1130 GBP
KPLI5120-0043
ABTS® Peroxidase Substrate Kit, 1-Component Microwell
Assays Enzyme Assays
KPL ABTS 1-component microwell peroxidase substrate develops a blue-green product when reacted with horseradish peroxidase labeled conjugates in microwell plates or tubes. The expected appearance of
the solution is colorless to light green. Acceptable background absorbance of unreacted substrate is ≤0,025, as measured at 405 nm. KPL ABTS 1-component microwell peroxidase substrate is not suitable for membrane or immunohistochemical staining.