Wash Solution Concentrate, 20X
Supplier: SERACARE LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
50-63-04
KPLI50-63-04EA 338 GBP
KPLI50-63-04
Wash Solution Concentrate, 20X
Buffers Electrophoresis Buffers Washing Buffers
Convenient 20X concentrate containing imidazole-buffered saline and Tween® 20.
  • Suitable for ELISA, blotting, and immunohistochemistry applications
This is used a general immunoassay wash buffer.