Heavy duty mobile tables
Supplier: KOETTERMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
507-00017 507-00009 507-00010 507-00011 507-00012 507-00013 507-00014 507-00016 507-00018 507-00019 507-00020 507-00021 507-00022 507-00023 507-00024 507-00025 507-00026 507-00027 507-00028 507-00015
KOTT507-00017EA 1670 GBP
KOTT507-00017 KOTT507-00009 KOTT507-00010 KOTT507-00011 KOTT507-00012 KOTT507-00013 KOTT507-00014 KOTT507-00016 KOTT507-00018 KOTT507-00019 KOTT507-00020 KOTT507-00021 KOTT507-00022 KOTT507-00023 KOTT507-00024 KOTT507-00025 KOTT507-00026 KOTT507-00027 KOTT507-00028 KOTT507-00015
Heavy duty mobile tables
Furniture Tables and Workbenches
Heavy duty mobile tables available with melamine TopResist® or laboratory grade SGL worktops.
Tables with both worktops suitable for:
Writing tasks
Chemical and equipment handling
Hygiene and aseptic requirement areas
Heavy duty mobile tables have sturdy castors, it can bear heavier weights.
Resistant to chemicals
Scratch resistance
Maximum loading capacity: 400 kg