Safety cabinets, poisons
Supplier: KOETTERMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
379-00003 379-00001 379-00000 379-00002
KOTT379-00003EA 333 GBP
KOTT379-00003 KOTT379-00001 KOTT379-00000 KOTT379-00002
Safety cabinets, poisons
Furniture Storage Cabinets
These cabinets are available with normal and hinged doors.
  • Lockable
  • Continuous extraction inside the cabinet
  • Can be wall mounted or installed in chemical storage cabinet
Epoxy powder coated steel.
Order Now
SPECIFICATIONS
Environmentally Preferable