Acid and alkali materials storage cabinets
Supplier: KOETTERMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
377-00008 377-00001 377-00002 377-00003 377-00004 377-00005 377-00007 377-00009 377-00010 377-00011 377-00012 377-00013 377-00014 377-00015 377-00016 377-00017 377-00018 377-00019 377-00020 377-00006
KOTT377-00008EA 2460 GBP
KOTT377-00008 KOTT377-00001 KOTT377-00002 KOTT377-00003 KOTT377-00004 KOTT377-00005 KOTT377-00007 KOTT377-00009 KOTT377-00010 KOTT377-00011 KOTT377-00012 KOTT377-00013 KOTT377-00014 KOTT377-00015 KOTT377-00016 KOTT377-00017 KOTT377-00018 KOTT377-00019 KOTT377-00020 KOTT377-00006
Acid and alkali materials storage cabinets
Furniture Storage Cabinets
These cabinets specially designed for the storage of highly corrosive products. It has separate compartments for storage of acids and bases. Liquid tight pull-out trays in the cabinet safely collect the spilled materials.
  • Optional integrated fan
  • Continuous extraction inside the cabinet
  • Cabinets without integrated fan should be connected to an extraction system
  • Maximum loading capacity: 30 kg per drawer
Polypropylene, acid resistant