Storage cabinets, tall

Supplier: KOETTERMANN
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

301-00001 301-00084 301-00064 301-00065 301-00066 301-00068 301-00069 301-00073 301-00075 301-00076 301-00077 301-00078 301-00079 301-00082 301-00058 301-00086 301-00089 301-00093 301-00095 301-00096 301-00097 301-00098 301-00080 301-00033 301-00002 301-00004 301-00005 301-00013 301-00015 301-00016 301-00017 301-00018 301-00019 301-00020 301-00026 301-00028 301-00062 301-00032 301-00060 301-00037 301-00039 301-00043 301-00045 301-00046 301-00047 301-00048 301-00050 301-00052 301-00054 301-00056 301-00030
KOTT301-00001EA 1730 GBP
KOTT301-00001 KOTT301-00084 KOTT301-00064 KOTT301-00065 KOTT301-00066 KOTT301-00068 KOTT301-00069 KOTT301-00073 KOTT301-00075 KOTT301-00076 KOTT301-00077 KOTT301-00078 KOTT301-00079 KOTT301-00082 KOTT301-00058 KOTT301-00086 KOTT301-00089 KOTT301-00093 KOTT301-00095 KOTT301-00096 KOTT301-00097 KOTT301-00098 KOTT301-00080 KOTT301-00033 KOTT301-00002 KOTT301-00004 KOTT301-00005 KOTT301-00013 KOTT301-00015 KOTT301-00016 KOTT301-00017 KOTT301-00018 KOTT301-00019 KOTT301-00020 KOTT301-00026 KOTT301-00028 KOTT301-00062 KOTT301-00032 KOTT301-00060 KOTT301-00037 KOTT301-00039 KOTT301-00043 KOTT301-00045 KOTT301-00046 KOTT301-00047 KOTT301-00048 KOTT301-00050 KOTT301-00052 KOTT301-00054 KOTT301-00056 KOTT301-00030
Storage cabinets, tall
Furniture Storage Cabinets
Tall cabinets provide especially large storage space.

  • Lockable doors
  • Colour: Arctic white

Galvanised epoxy powder coated steel.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR