Instrument dishes with lid, stainless steel
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2-1321
KOEH2-1321EA 141 GBP
KOEH2-1321
Instrument dishes with lid, stainless steel
Trays/Dishes/Pots Instrument Trays/Dishes
Stainless steel dishes with radiused corners, dull-bright finish and dust-tight knob lids.
  • Special corner shape for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance