Electron micrography films, Kodak
Supplier: CARESTREAM HEALTH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1679273 8971160 1608124 1615228 1493279
KODK1679273EA 0 GBP
KODK1679273 KODK8971160 KODK1608124 730-3213 KODK1493279
Electron micrography films, Kodak
Autoradiography Films Films
These micrography films offer considerable latitude in performance and flexibility for most electron microscopy applications. They are suitable for medium-energy electrons (40 to 100 ky) and acceptable results can also be obtained at higher electron levels.
  • Engineered to produce high-quality images
  • Transmission Electron Micrography (TEM) or electron diffraction studies
  • Versatile image capture in the transmission electron microscope for life and material research
Micrography Film 4489 with less curl and shorter pump-down times. Approximately half theMore Product Information