Cover glasses
Supplier: KNITTEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VD11818Y100A VD12440Y100A VD12432Y100A VD12550Y100A VD12460Y100A
KNITVD11818Y100AEA 75 GBP
KNITVD11818Y100A KNITVD12440Y100A KNITVD12432Y100A KNITVD12550Y100A KNITVD12460Y100A
Cover glasses
Cover Glasses
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
Certifications: ISO 8255/1
Packaging: 100 pieces per box - 1000 pieces per carton.