Dialysis tubing Spectra/Por®, biotech CE
Supplier: KLEINFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
131450 131450T
KLEI131450EA 1180 GBP
KLEI131450 KLEI131450T
Dialysis tubing Spectra/Por®, biotech CE
Tubing Dialysis Tubing
These cellulose Ester (CE) membrane offers the largest range of concise for desalting, isolating ionic species and macromolecular purifications which is supplied wet in 0,05 % or 0,1% sodium azide.
  • Superior chemical compatibility and temperature tolerance
  • Rigidly controlled porosity for better selectivity and separations
  • Ultrapure with no trace heavy metals & sulfides
  • Does not interfere with molecular activity and function
  • No glycerin removal or pretreatment required
Tubing is more sensitive to harsh conditions and solvents and generally toleratesMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable