Wipes for general applications, WYPALL* X70
Supplier: KIMBERLY CLARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8381 7867 8387 8389
115-1281EA 192 GBP
115-1281 115-1278 115-1282 KIMB8389
Wipes for general applications, WYPALL* X70
Wipes Reusable Cleaning Cloths Cloths for General Applications
Made from fast-absorbing HYDROKNIT* material, a hydroentangled cellulose/polypropylene blend without adhesives or chemical binders. For use in heavy manufacturing, metal fabrication, and processing plants for chemicals, refining, rubber, and plastics.
  • Durable, rag-tough construction and a textured surface, yet soft and pliable
  • Rags are reusable and will not fall apart when wet
  • Long lasting, on-the-job performance allows extended usage, reducing cost and waste
  • Large roll (115-2100) can be used with dispensers 115-0045, 115-1533 and 115-2038