Flange lids with side and center necks
Supplier: KGW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D4267 D4265 D4266 D4241
KGWAD4267EA 0 GBP
KGWAD4267 KGWAD4265 KGWAD4266 KGWAD4241
Flange lids with side and center necks
Lids Reaction Vessel Lids
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes