Cold traps, type KF NW
Supplier: KGW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17380-NW16
KGWA17380-NW16EA 1350 GBP
KGWA17380-NW16
Cold traps, type KF NW
Vacuum Traps
Simple cold traps, which are also known as cold finger condensers, are used in conjunction with vacuum pumps and the condensation Produced from humidity or solvents these cold traps can be used for many different tasks. With glass vacuum flanges you are able to connect this cold trap directly to the metal flanges of a high vacuum pump.
Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Reliable, perfect version, easy to clean and handle
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or
More Product Information