Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Supplier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N632433111014 N632433111016 632433111018 632433111019 N632433111022 632433111023 632433111025 632433111030 632433111037 632433119023 632433119025 632433119030
KAVAN632433111014EA 663 GBP
KAVAN632433111014 KAVAN632433111016 KAVA632433111018 KAVA632433111019 KAVAN632433111022 KAVA632433111023 KAVA632433111025 KAVA632433111030 KAVA632433111037 KAVA632433119023 KAVA632433119025 KAVA632433119030
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Pipettes Bulb Pipettes
Soda-lime glass.
  • Enlarged form
  • Non autoclavable at 121 °C
Certifications: DIN 12691, ISO 648