Funnel with short stem
Supplier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N632421001031 N632421001046 N632421001056 N632421001076 N632421001125 N632421001150 632413001300 N632421001030 N632421001041 N632421001050 N632421001060 N632421001071 N632421001081 N632421001086 N632421001100 632413001200 632413001250
221-1721EA 15.1 GBP
221-1721 221-1722 221-1723 221-1724 221-1727 221-1728 221-1731 KAVAN632421001030 KAVAN632421001041 KAVAN632421001050 KAVAN632421001060 KAVAN632421001071 KAVAN632421001081 KAVAN632421001086 KAVAN632421001100 KAVA632413001200 KAVA632413001250
Funnel with short stem
Funnels Filtration Funnels Filter Funnels
Borosilicate glass, grade 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With smooth internal walls.
Certifications: DIN 12445 and ISO 4798
Order Now
SPECIFICATIONS
Environmentally Preferable